siRNA Details
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009421.3 | NP_033447.2 | 6, 7 | 685 | ||
| GenBank mRNA | AK029135.1 | - | 6, 7 | 687 | ||
| AK089281.1 | - | 5, 6 | 429 | |||
| AK142931.1 | - | 5, 6 | 444 | |||
| AK145075.1 | - | 260 | ||||
| AK169515.1 | - | 6, 7 | 478 | |||
| AK172598.1 | - | 2, 3 | 289 | |||
| BC096595.1 | - | 2, 3 | 251 | 30748947 | ||
| L35302.1 | - | 5, 6 | 431 | |||

Traf1
Genomic Map