siRNA Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011643.2 | NP_035773.1 | 11, 12 | 2470 | ||
| GenBank mRNA | AF191551.1 | - | 10, 11 | 1916 | ||
| BC137581.1 | - | 11, 12 | 2456 | 8860600 | ||
| BC144726.1 | - | 10, 11 | 2354 | 9053274 | ||
| U40980.1 | - | 349 | ||||
| U73625.1 | - | 11, 12 | 2470 | |||
| U95167.1 | - | 10, 11 | 2368 | |||
| U95168.1 | - | 10, 11 | 2315 | |||
| U95169.1 | - | 9, 10 | 2213 | |||

Trpc1
Genomic Map