siRNA Details
| Gene Symbol: Trpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016984.3 | NP_058680.1 | 11 | 2575 | ||
| GenBank mRNA | AF011543.1 | - | 11 | 2392 | ||
| AK044265.1 | - | 11 | 2431 | |||
| AK080670.1 | - | 11 | 2576 | |||
| BC057335.1 | - | 11 | 2411 | |||
| BC060160.1 | - | 11 | 2411 | |||
| BC120729.1 | - | 11 | 2490 | 40129652 | ||
| BC131915.1 | - | 11 | 2490 | 40130192 | ||
| U50922.1 | - | 11 | 2528 | |||

Trpc4
Genomic Map