siRNA Details
| Gene Symbol: Tulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021478.1 | NP_067453.1 | 14 | 1403 | ||
| GenBank mRNA | AF085681.1 | - | 14 | 1451 | ||
| AF105711.1 | - | 14 | 1451 | |||
| AK053880.1 | - | 8 | 906 | |||
| AK164621.1 | - | 4 | 407 | |||
| BC014820.1 | - | 14 | 1409 | |||
| BC035952.1 | - | 1 | 1263 | |||
| BC038073.1 | - | 2 | 295 | |||

Tulp1
Genomic Map