siRNA Details
| Gene Symbol: Zrsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011663.3 | NP_035793.1 | 1 | 2118 | ||
| GenBank mRNA | AK030552.1 | - | 1 | 908 | ||
| BC132555.1 | - | 2 | 1176 | 40130832 | ||
| BC138594.1 | - | 1 | 1186 | 8861614 | ||
| D17407.1 | - | 2 | 2063 | |||
| S69507.1 | - | 1 | 2052 | |||

Zrsr1
Genomic Map