siRNA Details
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009458.4 | NP_033484.3 | 2, 3 | 263 | ||
GenBank mRNA | AK010432.1 | - | 2, 3 | 161 | ||
AK011363.1 | - | 2, 3 | 184 | |||
AK152316.1 | - | 2, 3 | 246 | |||
AK152645.1 | - | 2, 3 | 246 | |||
AK153174.1 | - | 2, 3 | 246 | |||
AK169229.1 | - | 2, 3 | 246 | |||
BC132075.1 | - | 2, 3 | 184 | 40130352 | ||
BC132077.1 | - | 2, 3 | 184 | 40130354 | ||
U57690.1 | - | 2, 3 | 176 | |||
X96859.1 | - | 2, 3 | 160 |
