siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136086.2 | NP_001129558.1 | 12 | 1505 | ||
| NM_001291455.1 | NP_001278384.1 | 12 | 1755 | |||
| NM_009468.5 | NP_033494.1 | 12 | 1686 | |||
| GenBank mRNA | AK077000.1 | - | 12 | 1496 | ||
| AK133040.1 | - | 12 | 1505 | |||
| AK149024.1 | - | 12 | 1523 | |||
| AK161509.1 | - | 12 | 1427 | |||
| AK171653.1 | - | 5 | 586 | |||
| BC023003.1 | - | 12 | 1370 | 5367381 | ||
| X87817.1 | - | 12 | 1646 | |||

Dpysl3
Genomic Map