siRNA Details
| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009474.5 | NP_033500.1 | 3 | 327 | ||
| GenBank mRNA | AK050047.1 | - | 2 | 117 | ||
| AK050425.1 | - | 3 | 328 | |||
| BC019771.1 | - | 3 | 309 | 5136328 | ||
| M27695.1 | - | 3 | 282 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK172422.1 | - | 40 | Chr. - - [-] on Build | ||

Uox
Genomic Map