siRNA Details
| Gene Symbol: Ggact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145466.2 | NP_663441.1 | 3 | 182 | ||
| GenBank mRNA | AK050251.1 | - | 3 | 172 | ||
| BC006662.1 | - | 3 | 152 | 3501534 | ||
| BC080839.1 | - | 138 | 30293286 | |||
| BY119987.1 | - | 3 | 180 | |||

Ggact
Genomic Map