siRNA Details
| Gene Symbol: Fam105a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242423.1 | NP_001229352.1 | 7 | 858 | ||
| NM_001242424.1 | NP_001229353.1 | 7 | 858 | |||
| NM_198301.2 | NP_938043.1 | 6 | 708 | |||
| GenBank mRNA | AK036522.1 | - | 7 | 833 | ||
| AK150760.1 | - | 7 | 829 | |||
| AK152120.1 | - | 7 | 829 | |||
| AK152590.1 | - | 7 | 824 | |||
| AK160030.1 | - | 7 | 802 | |||
| AK172362.1 | - | 7 | 812 | |||
| BC052328.1 | - | 6 | 664 | 6311697 | ||

Fam105a
Genomic Map