siRNA Details
| Gene Symbol: Slc32a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009508.2 | NP_033534.2 | 2 | 2005 | ||
| GenBank mRNA | AJ001598.1 | - | 2 | 1562 | ||
| AK140468.1 | - | 2 | 1766 | |||
| AK163661.1 | - | 2 | 1755 | |||
| AK163754.1 | - | 2 | 1781 | |||
| AY578289.1 | - | 2 | 2005 | |||
| BC052020.1 | - | 2 | 1996 | 5686843 | ||

Slc32a1
Genomic Map