siRNA Details
| Gene Symbol: Kcnk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033876.1 | NP_001029048.1 | 2 | 467 | ||
| GenBank mRNA | BC139167.1 | - | 2 | 528 | 8862189 | |
| BC139168.1 | - | 2 | 528 | 8862190 | ||
| DQ185133.1 | - | 2 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk9| Gene Symbol: Kcnk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033876.1 | NP_001029048.1 | 2 | 467 | ||
| GenBank mRNA | BC139167.1 | - | 2 | 528 | 8862189 | |
| BC139168.1 | - | 2 | 528 | 8862190 | ||
| DQ185133.1 | - | 2 | 467 | |||