siRNA Details
| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198607.1 | NP_941009.1 | 1 | 238 | ||
| GenBank mRNA | BC047266.1 | - | 1 | 238 | 6307114 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Them6| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198607.1 | NP_941009.1 | 1 | 238 | ||
| GenBank mRNA | BC047266.1 | - | 1 | 238 | 6307114 | |