siRNA Details
Gene Symbol: Nrbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144847.1 | NP_659096.1 | 8, 9 | 768 | ||
GenBank mRNA | BC011468.1 | - | 8, 9 | 701 | 4012880 | |
BC012437.1 | - | 8, 9 | 768 | 3710089 | ||
BC025042.1 | - | 3, 4 | 230 | |||
BC068117.1 | - | 13, 14 | 1416 |
