siRNA Details
| Gene Symbol: Gramd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172611.4 | NP_766199.1 | 17, 18 | 1758 | ||
| GenBank mRNA | AK036832.1 | - | 17, 18 | 1747 | ||
| AK129209.1 | - | 13, 14 | 3733 | |||
| AK142266.1 | - | 16, 17 | 2507 | |||
| BC021523.1 | - | 13, 14 | 1132 | |||
| BC139228.1 | - | 17, 18 | 1747 | 8862250 | ||
| BC139229.1 | - | 17, 18 | 1747 | 8862251 | ||

Gramd4
Genomic Map