siRNA Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145476.2 | NP_663451.2 | 9 | 1160 | ||
| GenBank mRNA | AK039627.1 | - | 9 | 1173 | ||
| AK146872.1 | - | 9 | 1161 | |||
| AK154226.1 | - | 9 | 1182 | |||
| AK154602.1 | - | 9 | 1203 | |||
| AK155459.1 | - | 9 | 1174 | |||
| AK155770.1 | - | 9 | 1203 | |||
| BC023106.1 | - | 6 | 555 | 4016550 | ||
| BC066009.1 | - | 9 | 1160 | 5708486 | ||

Tbc1d22a
Genomic Map