siRNA Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145476.2 | NP_663451.2 | 7 | 989 | ||
| GenBank mRNA | AK039627.1 | - | 7 | 1002 | ||
| AK146872.1 | - | 7 | 990 | |||
| AK154226.1 | - | 7 | 1011 | |||
| AK154602.1 | - | 7 | 1032 | |||
| AK155459.1 | - | 7 | 1003 | |||
| AK155770.1 | - | 7 | 1032 | |||
| AK216948.1 | - | 3 | 214 | |||
| BC066009.1 | - | 7 | 989 | 5708486 | ||

Tbc1d22a
Genomic Map