siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198927.3 | NP_945121.1 | 16 | 1864 | ||
| GenBank mRNA | AK132579.1 | - | 15 | 1767 | ||
| AY459348.1 | - | 16 | 1865 | |||
| BC022763.1 | - | 2 | 220 | |||
| BC038603.1 | - | 6 | 620 | |||
| BC049243.1 | - | 8 | 869 | |||
| BC049976.1 | - | 2 | 238 | |||
| BC055489.1 | - | 2 | 210 | |||
| BC069958.1 | - | 7 | 696 | |||
| BC138091.1 | - | 16 | 1865 | 8861110 | ||
| BC145136.1 | - | 11 | 1148 | 9053684 | ||

Smgc
Genomic Map