siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018765.3 | NP_061235.2 | 8 | 1071 | ||
| GenBank mRNA | AF071184.1 | - | 8 | 923 | ||
| AK083381.1 | - | 1 | 126 | |||
| AK135215.1 | - | 8 | 1071 | |||
| AK159522.1 | - | 8 | 1323 | |||
| AK160559.1 | - | 8 | 808 | |||
| AK161496.1 | - | 8 | 765 | |||
| AK162009.1 | - | 8 | 813 | |||
| AK164984.1 | - | 2 | 2304 | |||
| BC034851.1 | - | 8 | 766 | 1399984 | ||

Wbp4
Genomic Map