siRNA Details
Gene Symbol: Dbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207533.2 | NP_997416.2 | 2, 3 | 639 | ||
GenBank mRNA | AK035803.1 | - | 2, 3 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207533.2 | NP_997416.2 | 2, 3 | 639 | ||
GenBank mRNA | AK035803.1 | - | 2, 3 | 236 |