siRNA Details
| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172612.3 | NP_766200.1 | 1 | 202 | ||
| GenBank mRNA | AK039992.1 | - | 201 | |||
| AK043639.1 | - | 1 | 203 | |||
| AK161870.1 | - | 1 | 199 | |||
| BC048531.1 | - | 1 | 199 | 6535763 | ||

Rnd1
Genomic Map