siRNA Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 3 | 508 | ||
| GenBank mRNA | AF012923.1 | - | 3 | 462 | ||
| BC138740.1 | - | 2 | 327 | 8861762 | ||
| BC138742.1 | - | 2 | 327 | 8861764 | ||
| BQ895699.1 | - | 3 | 521 | |||
| CD804474.1 | - | 11 | ||||

Zmat3
Genomic Map