siRNA Details
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001983.2 | NP_001001983.2 | 18 | 2149 | ||
GenBank mRNA | BC075629.1 | - | 18 | 2149 | 6856662 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001983.2 | NP_001001983.2 | 18 | 2149 | ||
GenBank mRNA | BC075629.1 | - | 18 | 2149 | 6856662 |