siRNA Details
| Gene Symbol: Wnt10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009518.2 | NP_033544.1 | 3 | 656 | ||
| GenBank mRNA | AK013836.1 | - | 1 | 43 | ||
| BC014737.1 | - | 3 | 656 | 4921327 | ||
| U61969.1 | - | 1166 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt10a| Gene Symbol: Wnt10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009518.2 | NP_033544.1 | 3 | 656 | ||
| GenBank mRNA | AK013836.1 | - | 1 | 43 | ||
| BC014737.1 | - | 3 | 656 | 4921327 | ||
| U61969.1 | - | 1166 | ||||