siRNA Details
Gene Symbol: Wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271757.1 | NP_001258686.1 | 4, 5 | 857 | ||
NM_001271758.1 | NP_001258687.1 | 4, 5 | 800 | |||
NM_009525.3 | NP_033551.2 | 4, 5 | 1054 | |||
GenBank mRNA | AK049558.1 | - | 4, 5 | 824 | ||
BC010775.1 | - | 4, 5 | 800 | 4211437 | ||
BC051406.1 | - | 4, 5 | 985 | 6438917 | ||
BU520338.1 | - | 2, 3 | 466 | |||
M89799.1 | - | 3, 4 | 669 |
