siRNA Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271757.1 | NP_001258686.1 | 5 | 1084 | ||
| NM_001271758.1 | NP_001258687.1 | 5 | 1027 | |||
| NM_009525.3 | NP_033551.2 | 5 | 1281 | |||
| GenBank mRNA | AK013218.1 | - | 5 | 1252 | ||
| AK049558.1 | - | 5 | 1051 | |||
| BU520338.1 | - | 3 | 693 | |||
| M89799.1 | - | 4 | 896 | |||

Wnt5b
Genomic Map