siRNA Details
| Gene Symbol: Wnt7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009528.3 | NP_033554.3 | 1 | 387 | ||
| GenBank mRNA | BC052018.1 | - | 1 | 148 | 6401357 | |
| BC058398.1 | - | 1 | 387 | |||
| M89802.1 | - | 1 | 16 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt7b| Gene Symbol: Wnt7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009528.3 | NP_033554.3 | 1 | 387 | ||
| GenBank mRNA | BC052018.1 | - | 1 | 148 | 6401357 | |
| BC058398.1 | - | 1 | 387 | |||
| M89802.1 | - | 1 | 16 | |||