siRNA Details
| Gene Symbol: Wrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122822.1 | NP_001116294.1 | 33, 34 | 4181 | ||
| NM_011721.4 | NP_035851.3 | 33, 34 | 4291 | |||
| GenBank mRNA | AF091215.1 | - | 33, 34 | 4245 | ||
| AF241636.1 | - | 33, 34 | 4306 | |||
| AK052466.1 | - | 10, 11 | 1184 | |||
| AK161483.1 | - | 27, 28 | 4077 | |||
| BC050921.1 | - | 33, 34 | 4236 | 6408901 | ||
| BC060700.1 | - | 33, 34 | 4236 | 6408901 | ||
| D86526.1 | - | 33, 34 | 4261 | |||
| D86527.1 | - | 33, 34 | 4261 | |||

Wrn
Genomic Map