siRNA Details
| Gene Symbol: Xdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011723.3 | NP_035853.2 | 16 | 1741 | ||
| GenBank mRNA | AK034256.1 | - | 16 | 1689 | ||
| AK090058.1 | - | 7 | 882 | |||
| AK134156.1 | - | 2 | 226 | |||
| AK143412.1 | - | 16 | 1689 | |||
| AK144700.1 | - | 16 | 1693 | |||
| AK160010.1 | - | 13 | 1440 | |||
| AK161529.1 | - | 16 | 1692 | |||
| X62932.1 | - | 16 | 1675 | |||

Xdh
Genomic Map