siRNA Details
| Gene Symbol: Xirp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011724.3 | NP_035854.3 | 2 | 1212 | ||
| GenBank mRNA | AF051945.2 | - | 2 | 1183 | ||
| BC117959.1 | - | 2 | 1117 | 40098186 | ||
| BC117960.1 | - | 2 | 1117 | 40098187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xirp1| Gene Symbol: Xirp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011724.3 | NP_035854.3 | 2 | 1212 | ||
| GenBank mRNA | AF051945.2 | - | 2 | 1183 | ||
| BC117959.1 | - | 2 | 1117 | 40098186 | ||
| BC117960.1 | - | 2 | 1117 | 40098187 | ||