siRNA Details
| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145483.2 | NP_663458.2 | 5 | 594 | ||
| GenBank mRNA | AK143313.1 | - | 5 | 595 | ||
| AK173216.1 | - | 5 | 625 | |||
| BC015291.1 | - | 5 | 560 | 4242025 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp160| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145483.2 | NP_663458.2 | 5 | 594 | ||
| GenBank mRNA | AK143313.1 | - | 5 | 595 | ||
| AK173216.1 | - | 5 | 625 | |||
| BC015291.1 | - | 5 | 560 | 4242025 | ||