siRNA Details
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139154.2 | NP_631893.1 | 4 | 333 | ||
GenBank mRNA | AB232639.1 | - | 3 | 227 | ||
AF422144.1 | - | 3 | 286 | |||
AK133766.1 | - | 3 | 109 | |||
BC027008.1 | - | 4 | 321 | 5364768 | ||
BC037461.1 | - | 4 | 454 |
