siRNA Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146075.2 | NP_666187.2 | 8 | 1374 | ||
| GenBank mRNA | AK082131.1 | - | 8 | 1182 | ||
| AK143224.1 | - | 8 | 1359 | |||
| AY640307.1 | - | 8 | 1354 | |||
| BC026588.1 | - | 8 | 684 | 4972900 | ||
| BC050010.1 | - | 6 | 1612 | |||

Lemd2
Genomic Map