siRNA Details
| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172620.3 | NP_766208.2 | 4, 5 | 504 | ||
| GenBank mRNA | AK042520.1 | - | 4, 5 | 458 | ||
| AK051732.1 | - | 4, 5 | 575 | |||
| AK052523.1 | - | 4, 5 | 505 | |||
| AX775735.1 | - | 4, 5 | 316 | |||
| BC063329.1 | - | 4, 5 | 471 | 30251096 | ||
| KJ650214.1 | - | 4, 5 | 293 | |||

Vps52
Genomic Map