siRNA Details
| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172620.3 | NP_766208.2 | 8 | 922 | ||
| GenBank mRNA | AK042520.1 | - | 8 | 876 | ||
| AK051732.1 | - | 8 | 993 | |||
| AK052523.1 | - | 8 | 923 | |||
| AX775735.1 | - | 8 | 734 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps52| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172620.3 | NP_766208.2 | 8 | 922 | ||
| GenBank mRNA | AK042520.1 | - | 8 | 876 | ||
| AK051732.1 | - | 8 | 993 | |||
| AK052523.1 | - | 8 | 923 | |||
| AX775735.1 | - | 8 | 734 | |||