siRNA Details
| Gene Symbol: Ubr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177374.1 | NP_001170845.1 | 31 | 3814 | ||
| NM_146078.3 | NP_666190.2 | 31 | 3814 | |||
| GenBank mRNA | AK122254.2 | - | 29 | 4643 | ||
| AK143202.1 | - | 31 | 3814 | |||
| AK147445.1 | - | 31 | 3803 | |||
| AY061885.1 | - | 31 | 3501 | |||
| AY280958.1 | - | 31 | 3529 | |||
| BC026391.1 | - | 5 | 567 | |||
| BC031403.1 | - | 16 | 1608 | 4935220 | ||
| BC044903.1 | - | 17 | 1749 | |||
| BC075642.1 | - | 32 | 3628 | 30544183 | ||

Ubr2
Genomic Map