siRNA Details
| Gene Symbol: Prr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195673.1 | NP_001182602.1 | 3 | 600 | ||
| GenBank mRNA | BC172076.1 | - | 1 | 1135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr22| Gene Symbol: Prr22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195673.1 | NP_001182602.1 | 3 | 600 | ||
| GenBank mRNA | BC172076.1 | - | 1 | 1135 | ||