siRNA Details
| Gene Symbol: Dlgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128180.1 | NP_001121652.1 | 7 | 1619 | ||
| NM_001128181.1 | NP_001121653.1 | 8 | 1621 | |||
| NM_027712.3 | NP_081988.3 | 8 | 2606 | |||
| NM_177639.6 | NP_808307.2 | 9 | 2636 | |||
| GenBank mRNA | AK034182.1 | - | 3 | 495 | ||
| AK035763.1 | - | 12 | 3187 | |||
| AK136926.1 | - | 8 | 1582 | |||
| AK147448.1 | - | 12 | 3423 | |||
| AK161941.1 | - | 7 | 1594 | |||
| AK163689.1 | - | 7 | 1600 | |||
| AK220515.1 | - | 11 | 3486 | |||
| AY243846.2 | - | 8 | 2387 | |||
| BC062120.1 | - | 9 | 2637 | 6822047 | ||
| BC094369.1 | - | 7 | 1815 | |||

Dlgap1
Genomic Map