siRNA Details
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 1, 2 | 227 | ||
GenBank mRNA | AI430883.1 | - | 1, 2 | 227 | ||
AK014711.1 | - | 1, 2 | 216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 1, 2 | 227 | ||
GenBank mRNA | AI430883.1 | - | 1, 2 | 227 | ||
AK014711.1 | - | 1, 2 | 216 |