siRNA Details
| Gene Symbol: Mib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144860.2 | NP_659109.2 | 17 | 3114 | ||
| GenBank mRNA | AK053035.1 | - | 10 | 1337 | ||
| AK080847.1 | - | 11 | 1390 | |||
| AK161848.1 | - | 2 | 210 | |||
| AY147848.1 | - | 17 | 2613 | |||
| AY149907.1 | - | 17 | 2434 | |||
| AY245539.1 | - | 17 | 3114 | |||
| AY974091.1 | - | 17 | 2407 | |||
| BC011287.1 | - | 6 | 711 | |||
| BC083072.1 | - | 7 | 792 | |||
| BC131662.1 | - | 18 | 2580 | 40110696 | ||

Mib1
Genomic Map