siRNA Details
Gene Symbol: Hrh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153087.2 | NP_694727.1 | 1 | 199 | ||
GenBank mRNA | AF358859.1 | - | 1 | 199 | ||
BC111862.1 | - | 1 | 198 | 40046148 | ||
EU660534.1 | - | 1 | 152 | |||
EU660535.1 | - | 1 | 172 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
