siRNA Details
| Gene Symbol: Rbm27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172626.2 | NP_766214.2 | 16 | 2591 | ||
| GenBank mRNA | AK087759.1 | - | 3 | 171 | ||
| AK129330.1 | - | 11 | 1600 | |||
| AY461716.1 | - | 16 | 2566 | |||
| BC054080.1 | - | 12 | 1907 | 6390990 | ||

Rbm27
Genomic Map