siRNA Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172627.3 | NP_766215.1 | 8 | 915 | ||
| GenBank mRNA | AK028188.1 | - | 8 | 774 | ||
| AK081731.1 | - | 8 | 916 | |||
| BC048404.1 | - | 3 | 262 | |||
| BC141021.1 | - | 8 | 913 | 9055633 | ||

Pggt1b
Genomic Map