siRNA Details
| Gene Symbol: Fam170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004061.1 | NP_001004061.1 | 2 | 398 | ||
| GenBank mRNA | AY692449.1 | - | 2 | 398 | ||
| BC147592.1 | - | 2 | 396 | 9056582 | ||
| BC147595.1 | - | 2 | 396 | 9056585 | ||
| BC172108.1 | - | 2 | 396 | |||

Fam170a
Genomic Map