siRNA Details
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033281.3 | NP_001028453.1 | 4 | 1067 | ||
GenBank mRNA | AK133910.1 | - | 4 | 1068 | ||
BC139295.1 | - | 4 | 1097 | 8862317 | ||
DQ340564.1 | - | 4 | 643 | |||
DQ340565.1 | - | 4 | 562 |
