siRNA Details
| Gene Symbol: Slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081072.1 | NP_001074541.1 | 9 | 1802 | ||
| GenBank mRNA | AK028699.1 | - | 9 | 1804 | ||
| BC115435.1 | - | 9 | 1785 | 40094587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc27a6| Gene Symbol: Slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081072.1 | NP_001074541.1 | 9 | 1802 | ||
| GenBank mRNA | AK028699.1 | - | 9 | 1804 | ||
| BC115435.1 | - | 9 | 1785 | 40094587 | ||