siRNA Details
| Gene Symbol: Onecut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194268.2 | NP_919244.2 | 1 | 41 | ||
| GenBank mRNA | AY242995.1 | - | 1 | 41 | ||
| BC103668.1 | - | 1 | 18 | 40046651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Onecut2| Gene Symbol: Onecut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194268.2 | NP_919244.2 | 1 | 41 | ||
| GenBank mRNA | AY242995.1 | - | 1 | 41 | ||
| BC103668.1 | - | 1 | 18 | 40046651 | ||