siRNA Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146089.2 | NP_666201.1 | 9 | 833 | ||
| GenBank mRNA | AK076912.1 | - | 9 | 813 | ||
| AK146923.1 | - | 9 | 830 | |||
| AK147599.1 | - | 3 | 6714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Haus1| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146089.2 | NP_666201.1 | 9 | 833 | ||
| GenBank mRNA | AK076912.1 | - | 9 | 813 | ||
| AK146923.1 | - | 9 | 830 | |||
| AK147599.1 | - | 3 | 6714 | |||