siRNA Details
| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024560.2 | NP_001019731.1 | 9, 10 | 934 | ||
| GenBank mRNA | AK047234.1 | - | 9, 10 | 910 | ||
| BC048922.1 | - | 9, 10 | 934 | 6311864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx32| Gene Symbol: Snx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024560.2 | NP_001019731.1 | 9, 10 | 934 | ||
| GenBank mRNA | AK047234.1 | - | 9, 10 | 910 | ||
| BC048922.1 | - | 9, 10 | 934 | 6311864 | ||