siRNA Details
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001984.2 | NP_001001984.2 | 19 | 3865 | ||
GenBank mRNA | AK141779.1 | - | 5 | 1106 | ||
AK173084.1 | - | 8 | 1683 | |||
BC036264.1 | - | 7 | 1564 | |||
BC037117.1 | - | 12 | 2414 | |||
BC057051.1 | - | 6 | 1238 | |||
BC076576.1 | - | 19 | 3430 | 6840591 |
